missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM29 Antibody (TRIM29/9256R), Novus Biologicals™
Rabbit Monoclonal Antibody
268.00 € - 496.00 €
Specifications
| Antigen | TRIM29 |
|---|---|
| Clone | TRIM29/9256R |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin : 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18661795
|
Novus Biologicals
NBP3-23806-20ug |
20 μg |
268.00 €
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18680924
|
Novus Biologicals
NBP3-23806-100ug |
100 μg |
496.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRIM29 Monoclonal antibody specifically detects TRIM29 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| TRIM29 | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Ataxia telangiectasia group D-associated protein, ATDC, ATDC ataxia-telangiectasia group D-associated protein, FLJ36085, tripartite motif containing 29, tripartite motif protein TRIM29, tripartite motif-containing 29, tripartite motif-containing protein 29 | |
| Recombinant fragment (around aa 1-200) of human TRIM29 protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
| TRIM29/9256R | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| 1 mM PBS with 0.05% BSA | |
| 23650 | |
| IgG κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title