missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TRIM31 Polyclonal antibody specifically detects TRIM31 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | TRIM31 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 |
| Formulation | PBS (pH 7.3), 50% glycerol |
| Gene Alias | C6orf13E3 ubiquitin-protein ligase TRIM31, EC 6.3.2.-, HCG1, HCGItripartite motif-containing 31 gamma, RNFring finger protein, tripartite motif containing 31, tripartite motif-containing 31, Tripartite motif-containing protein 31 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 186-425 of human TRIM31 (NP_008959.3). ELLHQVLEEEKNFLLSRIYWLGHEGTEAGKHYVASTEPQLNDLKKLVDSLKTKQNMPPRQLLEDIKVVLCRSEEFQFLNPTPVPLELEKKLSEAKSRHDSITGSLKKFKDQLQADRKKDENRFFKSMNKNDMKSWGLLQKNNHKMNKTSEPGSSSAGGRTTSGPPNHHSSAPSHSLFRASSAGKVTFPVCLLASYDEISGQGASSQDTKTFDVALSEELHAALSEWLTAIRAWFCEVPSS |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?