missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM60 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94453-0.02ml
This item is not returnable.
View return policy
Description
TRIM60 Polyclonal antibody specifically detects TRIM60 in Mouse samples. It is validated for Western Blot
Specifications
| TRIM60 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| FLJ35882, RING finger protein 129MGC119325, RING finger protein 33, RNF129, RNF33, tripartite motif containing 60, tripartite motif-containing 60, tripartite motif-containing protein 60 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human TRIM60 (NP_689833.1). MEFVTALVNLQEESSCPICLEYLKDPVTINCGHNFCRSCLSVSWKDLDDTFPCPVCRFCFPYKSFRRNPQLRNLTEIAKQLQIRRSKRKRQKENAMCEKHNQFLTLFCVKDLEILCTQCSFSTKHQKHYICPIKKAASYHREILEGSLEPLRNNIERVEKVIILQGSKSVELKKKVEYKREEINSEFEQIRLFLQNEQEMILRQIQDEEMNILAKLNENLVELSDYVSTLKHLLREVEGKSVQSNLELLTQAKSMHHKYQNLKCPELFSFRLTKYGFSLPPQYSGLDRIIKPFQVDVILD | |
| 0.02 mL | |
| Zinc Finger | |
| 166655 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction