missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM60 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00 € - 470.00 €
Specifications
| Antigen | TRIM60 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|
18605102
|
Novus Biologicals
NBP2-94453-0.02ml |
0.02 mL |
190.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18623662
|
Novus Biologicals
NBP2-94453-0.1ml |
0.1 mL |
470.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
TRIM60 Polyclonal antibody specifically detects TRIM60 in Mouse samples. It is validated for Western BlotSpezifikation
| TRIM60 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Zinc Finger | |
| PBS (pH 7.3), 50% glycerol | |
| 166655 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse | |
| FLJ35882, RING finger protein 129MGC119325, RING finger protein 33, RNF129, RNF33, tripartite motif containing 60, tripartite motif-containing 60, tripartite motif-containing protein 60 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human TRIM60 (NP_689833.1). MEFVTALVNLQEESSCPICLEYLKDPVTINCGHNFCRSCLSVSWKDLDDTFPCPVCRFCFPYKSFRRNPQLRNLTEIAKQLQIRRSKRKRQKENAMCEKHNQFLTLFCVKDLEILCTQCSFSTKHQKHYICPIKKAASYHREILEGSLEPLRNNIERVEKVIILQGSKSVELKKKVEYKREEINSEFEQIRLFLQNEQEMILRQIQDEEMNILAKLNENLVELSDYVSTLKHLLREVEGKSVQSNLELLTQAKSMHHKYQNLKCPELFSFRLTKYGFSLPPQYSGLDRIIKPFQVDVILD | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts