missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimipramine 1.0 mg/ml in Methanol, LoGiCal
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Description
Trimipramine 1.0 mg/ml in Methanol
Specifications
Specifications
| Chemical Name or Material | Trimipramine |
| CAS | 739-71-9 |
| Analyte or Component Names | Trimipramine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H26 N2 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Trimipramine |
| InChI Formula | InChI=1 S/C20H26N2/c1-16(14-21(2)3)15-22-19-10-6-4-8-17(19)12-13-18-9-5-7-11-20(18)22/h4-11,16 H,12-15H2,1-3H3 |
| SMILES | CC(CN(C)C)CN1c2ccccc2CCc3ccccc13 |
| IUPAC Name | 3-(5,6-dihydrobenzo[b][1 ]benzazepin-11-yl)-N,N,2-trimethylpropan-1-amine |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction