missing translation for 'onlineSavingsMsg'
Learn More

Trimipramine Maleate 1.0 mg/ml in Methanol (as free base), LoGiCal

Product Code. 30463571
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30463571 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30463571 Supplier LoGiCal Supplier No. LGCAMP0240.0001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

Trimipramine Maleate 1.0 mg/ml in Methanol (as free base)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Trimipramine maleate
CAS 521-78-8
Analyte or Component Names Trimipramine maleate
Shipping Condition Ice Pack (-20°C)
Molecular Formula C20 H26 N2 . C4 H4 O4
Certifications/Compliance ISO 17034
Synonym Trimipramine Maleate
InChI Formula InChI=1 S/C20H26N2.C4H4O4/c1-16(14-21(2)3)15-22-19-10-6-4-8-17(19)12-13-18-9-5-7-11-20(18)22;5-3(6)1-2-4(7)8/h4-11,16 H,12-15H2,1-3H3;1-2 H,(H,5,6)(H,7,8)/b;2-1-
SMILES CC(CN(C)C)CN1c2ccccc2CCc3ccccc13.OC(=O)\C=C/C(=O)O
IUPAC Name (Z)-but-2-enedioic acid;3-(5,6-dihydrobenzo[b][1 ]benzazepin-11-yl)-N,N,2-trimethylpropan-1-amine
Solvent or Matrix Methanol
Industry Type Forensics
Molecular Weight (g/mol) 410.51
Quantity 1 mL
Formula Weight 410.2206 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.