missing translation for 'onlineSavingsMsg'
Learn More

Triphenyl phosphate (TPP) 500 μg/mL in Methyl tert-butyl ether, Dr. Ehrenstorfer

Product Code. 30311144
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30311144 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30311144 Supplier Dr. Ehrenstorfer Supplier No. DREGA09011131MB
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Triphenylphosphate
CAS 115-86-6
Analyte or Component Names Triphenylphosphate
Shipping Condition Ice pack (-20°C)
Molecular Formula C18 H15 O4 P
Concentration 500 μg/mL
Certifications/Compliance ISO 17034
Synonym Phosphoric acid, triphenyl ester, Celluflex TPP, DHPF 005, Disflamoll TP, NSC 57868, Phenyl phosphate ((PhO)3PO), Phoscon FR 903N, Phosflex TPP, Reofos TPP, S 4, S 4 (phosphate), Sumilizer TPP, TFF, TP, TPP, TPPA, TTP, TTP (triphenyl phosphate), Triphenol phosphate, Triphenoxyphosphine oxide, Triphenyl phosphate, WSFR-TPP, Wako TPP
InChI Formula InChI=1S/C18H15O4P/c19-23(20-16-10-4-1-5-11-16,21-17-12-6-2-7-13-17)22-18-14-8-3-9-15-18/h1-15H
SMILES O=P(Oc1ccccc1)(Oc2ccccc2)Oc3ccccc3
IUPAC Name triphenyl phosphate
Solvent or Matrix MTBE
Molecular Weight (g/mol) 326.28
Quantity 1 mL
Formula Weight 326.0708 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.