missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Triphenylacrylonitrile, 98+%

Product Code. p-7043532
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11374484 5 g 5g
11384484 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11374484 Supplier Alfa Aesar™ Supplier No. L01158.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 6304-33-2
Molecular Formula C21H15N
Molecular Weight (g/mol) 281.36
MDL Number MFCD00013811
InChI Key OPACMJHTQWVSKW-UHFFFAOYSA-N
Synonym triphenylacrylonitrile, 2,3,3-triphenylacrylonitrile, triphenylcyanoethylene, acrylonitrile, triphenyl, 2,3,3-triphenyl-acrylonitrile, acrylonitrile, 2,3,3-triphenyl, alpha,beta-diphenylcinnamonitrile, 2,3,3-trifenylakrylonitril czech, alpha-diphenylmethylene benzeneacetic acid, benzeneacetic acid, alpha-diphenylmethylene
PubChem CID 22743
ChEBI CHEBI:34217
IUPAC Name 2,3,3-triphenylprop-2-enenitrile
SMILES N#CC(=C(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 165°C to 167°C
Quantity 5 g
UN Number UN3439
Beilstein 1980289
Formula Weight 281.36
Percent Purity ≥98%
Chemical Name or Material Triphenylacrylonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.