missing translation for 'onlineSavingsMsg'
Learn More

Triphenylphosphine Oxide, Mikromol™

Product Code. 17941416
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17941416 250 mg 250mg
30052089 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17941416 Supplier Mikromol Supplier No. MM1219.100250
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Triphenylphosphine Oxide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Triphenylphosphine oxide
CAS 791-28-6
Analyte or Component Names Triphenylphosphine oxide
Shipping Condition Room Temperature
Molecular Formula C18 H15 O P
Certifications/Compliance ISO 17034
Synonym Triphenylphosphine oxide, Triphenyl phosphine oxide, Orlistat Related Compound C, Alprostadil Impurity K
InChI Formula InChI=1S/C18H15OP/c19-20(16-10-4-1-5-11-16,17-12-6-2-7-13-17)18-14-8-3-9-15-18/h1-15H
SMILES O=P(c1ccccc1)(c2ccccc2)c3ccccc3
IUPAC Name diphenylphosphorylbenzene
Molecular Weight (g/mol) 278.28
Quantity 250 mg
Formula Weight 278.0861 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.