missing translation for 'onlineSavingsMsg'
Learn More

(4-Chlorophenylsulfonyl)acetic acid, 98%, Thermo Scientific™

Product Code. 11387229
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 11387229

Brand: Thermo Scientific Alfa Aesar H27414.MC

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3405-89-8
Molecular Formula C8H7ClO4S
Molecular Weight (g/mol) 234.65
MDL Number MFCD00201948
InChI Key IAONCYGNBVHNCT-UHFFFAOYSA-N
Synonym 2-4-chlorophenyl sulfonyl acetic acid, 4-chlorophenylsulfonylacetic acid, 4-chlorophenyl sulfonyl acetic acid, 4-chloro-benzenesulfonyl-acetic acid, 2-4-chlorobenzenesulfonyl acetic acid, 4-chlorophenylsulfonyl acetic acid, 4-chlorobenzenesulfonyl acetic acid, cambridge id 5493053, chlorophenylsulfonylaceticacid
PubChem CID 55680
IUPAC Name 2-(4-chlorophenyl)sulfonylacetic acid
SMILES C1=CC(=CC=C1S(=O)(=O)CC(=O)O)Cl

Specifications

Melting Point ∽140°C (decomposition)
Quantity 100 mg
Formula Weight 234.66
Percent Purity 98%
Chemical Name or Material (4-Chlorophenylsulfonyl)acetic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.