missing translation for 'onlineSavingsMsg'
Learn More

TrkC Antibody Pair (HRP), Novus Biologicals™

Product Code. 18225837 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
15 Plates
Unit Size:
Pack of 15
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18225837 15 Plates Pack of 15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18225837 Supplier Novus Biologicals Supplier No. NBP27884715PLATES
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TrkC Monoclonal specifically detects TrkC in Human samples. It is validated for ELISA/Sandwich ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TrkC
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Conjugate HRP
Dilution Sandwich ELISA
Gene Alias EC 2.7.10, EC 2.7.10.1, ETS related protein-neurotrophic receptor tyrosine kinase fusion protein, ETV6-NTRK3 fusion, gp145(trkC), GP145-TrkC, Neurotrophic tyrosine kinase receptor type 3, neurotrophic tyrosine kinase, receptor, type 3, NT 3 receptor, NT-3 growth factor receptor, Trk-C, TrkC tyrosine kinase, TRKCneurotrophin 3 receptor, tyrosine kinase receptor C
Gene Symbols NTRK3
Host Species Mouse
Quantity 15 Plates
Research Discipline Cancer, Phospho Specific, Protein Kinase
Gene ID (Entrez) 4916
Test Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human TrkC.
Target Species Human
Content And Storage Storage of components varies. See protocol for specific instructions.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.