missing translation for 'onlineSavingsMsg'
Learn More

Trophinin Antibody - BSA Free, Novus Biologicals™

Product Code. 18178527 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
Unit Size:
0.03mg
0.1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18178527 0.1 mg 0.1mg
18675876 0.025 mg 0.03mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18178527 Supplier Novus Biologicals Supplier No. NBP241320

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Trophinin Polyclonal specifically detects Trophinin in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Trophinin
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 μg/mL, ELISA, Immunohistochemistry 10 μg/mL, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 10 μg/mL
Formulation PBS with 0.02% Sodium Azide
Gene Alias KIAA1114, MAGE Superfamily Protein, MAGE-d3, MAGED3, MAGE-D3 Antigen, magphinin, Melanoma Antigen, Family D, 3, trophinin
Gene Symbols TRO
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid peptide near the amino terminus of human Trophinin. The immunogen is located within amino acids 390 - 440 of Trophinin. Amino Acid Squence: EPTTRTRGKRNRKSKH
Molecular Weight of Antigen 94 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7216
Test Specificity Multiple isoforms of Trophinin are known to exist.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.