missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRPC1 Recombinant Rabbit Monoclonal Antibody (24GB4145)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557294
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
TRPC1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Thought to form a receptor-activated non-selective calcium permeant cation channel. Probably is operated by a phosphatidylinositol second messenger system activated by receptor tyrosine kinases or G-protein coupled receptors. Seems to be also activated by intracellular calcium store depletion. Interacts with ITPR3. Appears to be ubiquitous.
Spezifikation
| TRPC1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Trpc1 | |
| capacitative calcium channel protein Trp1; capacitative calcium entry channel; HTRP-1; Mtrp1; Short transient receptor potential channel 1; transient receptor potential canonical 1; transient receptor potential cation channel subfamily C member 1; transient receptor potential cation channel, subfamily C, member 1; transient receptor potential cation channel, subfamily C, member 1 L homeolog; transient receptor potential protein; transient receptor protein 1; trp; TRP1; TRP-1; trpc1; trpc1.L; trp-related protein 1; Trrp1; XELAEV_18027727mg; xtrpc1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 22063, 7220, 89821 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB4145 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P48995, Q61056, Q9QX01 | |
| Trpc1 | |
| A synthesized peptide derived from human TRPC1 (aa 411-492). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur