missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRPC1 Recombinant Rabbit Monoclonal Antibody (24GB4145)

Rabbit Recombinant Monoclonal Antibody
146.00 € - 500.00 €
Specifications
| Antigen | TRPC1 |
|---|---|
| Clone | 24GB4145 |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30276345
![]() |
Invitrogen™
MA557293 |
100 μL |
500.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30274874
|
Invitrogen™
MA557294 |
20 μL |
146.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRPC1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Thought to form a receptor-activated non-selective calcium permeant cation channel. Probably is operated by a phosphatidylinositol second messenger system activated by receptor tyrosine kinases or G-protein coupled receptors. Seems to be also activated by intracellular calcium store depletion. Interacts with ITPR3. Appears to be ubiquitous.Specifications
| TRPC1 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| Trpc1 | |
| capacitative calcium channel protein Trp1; capacitative calcium entry channel; HTRP-1; Mtrp1; Short transient receptor potential channel 1; transient receptor potential canonical 1; transient receptor potential cation channel subfamily C member 1; transient receptor potential cation channel, subfamily C, member 1; transient receptor potential cation channel, subfamily C, member 1 L homeolog; transient receptor potential protein; transient receptor protein 1; trp; TRP1; TRP-1; trpc1; trpc1.L; trp-related protein 1; Trrp1; XELAEV_18027727mg; xtrpc1 | |
| Trpc1 | |
| Primary | |
| Affinity chromatography |
| 24GB4145 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P48995, Q61056, Q9QX01 | |
| 22063, 7220, 89821 | |
| A synthesized peptide derived from human TRPC1 (aa 411-492). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title