Learn More
Invitrogen™ TRPC1 Recombinant Rabbit Monoclonal Antibody (24GB4145)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557293
 View more versions of this product
Product Code. 30276345
  • 500.00 € / 100µL
Description
TRPC1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Thought to form a receptor-activated non-selective calcium permeant cation channel. Probably is operated by a phosphatidylinositol second messenger system activated by receptor tyrosine kinases or G-protein coupled receptors. Seems to be also activated by intracellular calcium store depletion. Interacts with ITPR3. Appears to be ubiquitous.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TRPC1
Recombinant Monoclonal
Unconjugated
Trpc1
capacitative calcium channel protein Trp1; capacitative calcium entry channel; HTRP-1; Mtrp1; Short transient receptor potential channel 1; transient receptor potential canonical 1; transient receptor potential cation channel subfamily C member 1; transient receptor potential cation channel, subfamily C, member 1; transient receptor potential cation channel, subfamily C, member 1 L homeolog; transient receptor potential protein; transient receptor protein 1; trp; TRP1; TRP-1; trpc1; trpc1.L; trp-related protein 1; Trrp1; XELAEV_18027727mg; xtrpc1
Rabbit
Affinity chromatography
RUO
22063, 7220, 89821
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB4145
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P48995, Q61056, Q9QX01
Trpc1
A synthesized peptide derived from human TRPC1 (aa 411-492).
100 μL
Primary
Human, Mouse, Rat