missing translation for 'onlineSavingsMsg'
Learn More

TRUSS Antibody (1C0) [Alexa Fluor« 594], Novus Biologicals Biologicals™

Product Code. 30519551 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30519551 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30519551 Supplier Novus Biologicals Supplier No. NBP343055AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TRUSS Monoclonal antibody specifically detects TRUSS in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRUSS
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 1C0
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias C20orf188, dJ756N5.2, DKFZp586C1223, DKFZP727M231, Protein TAP1, Protein TRUSS, short transient receptor potential channel 4 associated protein, short transient receptor potential channel 4-associated protein, TNF-receptor ubiquitous scaffolding/signaling protein, transient receptor potential cation channel, subfamily C, member 4 associatedprotein, Trp4-associated protein, Trpc4-associated protein, TRRP4APchromosome 20 open reading frame 188, TRUSS, tumor necrosis factor receptor-associated ubiquitous scaffolding and signalingprotein
Host Species Mouse
Immunogen transient receptor potential cation channel, subfamily C, member 4 associated protein - (Uniprot# Q8TEL6)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26133
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.