missing translation for 'onlineSavingsMsg'
Learn More

Tryptophan, Mikromol™

Product Code. 30459328
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30459328 4 x 25 mg 100mg
30051458 25 mg 25mg
17970035 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30459328 Supplier Mikromol Supplier No. MM1267.0100254AMP
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Tryptophan, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DL-Tryptophan
CAS 54-12-6
Analyte or Component Names DL-Tryptophan
Shipping Condition Room Temperature
Molecular Formula C11 H12 N2 O2
Certifications/Compliance ISO 17025
Synonym Tryptophan, N-Acetyltryptophan Imp. A (EP), N-Acetyltryptophan Impurity A
InChI Formula InChI=1S/C11H12N2O2/c12-9(11(14)15)5-7-6-13-10-4-2-1-3-8(7)10/h1-4,6,9,13H,5,12H2,(H,14,15)
Solution Type Neat
SMILES NC(Cc1c[nH]c2ccccc12)C(=O)O
IUPAC Name 2-amino-3-(1H-indol-3-yl)propanoic acid
Molecular Weight (g/mol) 204.23
Quantity 4 x 25 mg
Formula Weight 204.0899 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.