missing translation for 'onlineSavingsMsg'
Learn More

TTF2 Antibody [mFluor Violet 610 SE], Novus Biologicals Biologicals™

Product Code. 30513541 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30513541 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30513541 Supplier Novus Biologicals Supplier No. NBP338460MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TTF2 Polyclonal antibody specifically detects TTF2 in Human samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTF2
Applications ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Formulation 50mM Sodium Borate
Gene Alias EC 3.6.1, EC 3.6.4.-, F2, HuF2lodestar protein, human factor 2, Lodestar homolog, RNA polymerase II termination factor, Transcription release factor 2, transcription termination factor 2, transcription termination factor, RNA polymerase II
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human TTF2 (NP_003585.3).,, Sequence:, MEEVRCPEHGTFCFLKTGVRDGPNKGKSFYVCRADTCSFVRATDIPVSHCLLHEDFVVELQGLLLPQDKKEYRLFFRCIRSKAEGKRWCGSIPWQDPDSKEHSVSNKSQHASETFHHSSNWLRNPFKVLDKNQEPALWKQLIKGEGEEKKADKKQREKGDQLFDQKKEQKPEMMEKDLSSGLVPKKKQSVVQEKKQEEGAEIQCEAETGGTHKRDFSEIKSQQCQGNELTRPSASSQEKSSGKSQDVQRESEPLREKVTQLLPQNVHSHN
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8458
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.