missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TWEAK/TNFSF12 Antibody (MTW-1) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
250.00 € - 705.00 €
Specifications
| Antigen | TWEAK/TNFSF12 |
|---|---|
| Clone | MTW-1 |
| Concentration | 1 mg/mL |
| Applications | Flow Cytometry, Immunofluorescence, Neutralization |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18693758
|
Novus Biologicals
NBP3-09045-0.025mg |
0.025 mg |
250.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18696797
|
Novus Biologicals
NBP3-09045-0.2mg |
0.2 mg |
705.00 €
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TWEAK/TNFSF12 Monoclonal antibody specifically detects TWEAK/TNFSF12 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, NeutralizationSpecifications
| TWEAK/TNFSF12 | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| APO3 ligand, APO3LMGC20669, DR3LGTWEAKAPO3/DR3 ligand, MGC129581, TNF-related weak inducer of apoptosis, tumor necrosis factor (ligand) superfamily, member 12, tumor necrosis factor ligand superfamily member 12 | |
| This antibody was raised by immunising SD rats with CD8-mTWEAK, a fusion of the human CD8alpha extracellular region to murine TWEAK. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| MTW-1 | |
| Flow Cytometry, Immunofluorescence, Neutralization | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Apoptosis, Cancer, Tumor Suppressors | |
| PBS | |
| 8742 | |
| IgG κ | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title