missing translation for 'onlineSavingsMsg'
Learn More
Learn More
u-Plasminogen Activator/Urokinase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-21338-25ul
This item is not returnable.
View return policy
Description
u-Plasminogen Activator/Urokinase Polyclonal antibody specifically detects u-Plasminogen Activator/Urokinase in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| u-Plasminogen Activator/Urokinase | |
| Polyclonal | |
| Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| ATF, EC 3.4.21, EC 3.4.21.73, plasminogen activator, urokinase, uPA, u-PA, U-plasminogen activator, urinary, urokinase-type plasminogen activator | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: KPSSPPEELKFQCGQKTLRPRFKIIGGEFTTIENQPWFAAIYRRHRGGSVTYVCGGSLISPCWVISATHCFIDYP | |
| 25 μg | |
| Cancer | |
| 5328 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH 7.2, 40% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction