missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UbcH2/UBE2H Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
227.00 € - 326.00 €
Specifications
| Antigen | UbcH2/UBE2H |
|---|---|
| Dilution | Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunoprecipitation 0.5-2 uL/mg of lysate |
| Applications | Western Blot, ELISA, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18695525
|
Novus Biologicals
NBP3-00048-100ul |
100 μg |
326.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18653586
|
Novus Biologicals
NBP3-00048-50ul |
50 μg |
227.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
UbcH2/UBE2H Polyclonal antibody specifically detects UbcH2/UBE2H in Human samples. It is validated for Western Blot, ELISA, ImmunoprecipitationSpecifications
| UbcH2/UBE2H | |
| Western Blot, ELISA, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Cell Biology | |
| 0.2 um filtered solution in PBS | |
| 7328 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunoprecipitation 0.5-2 uL/mg of lysate | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 6.3.2.19, UBC8E2-20K, UBCH, UBCH2, Ubiquitin carrier protein H, ubiquitin-conjugating enzyme E2 H, Ubiquitin-conjugating enzyme E2-20K, ubiquitin-conjugating enzyme E2H (homologous to yeast UBC8), ubiquitin-conjugating enzyme E2H (UBC8 homolog, yeast), Ubiquitin-protein ligase H | |
| Produced in rabbits immunized with purified, recombinant Human UbcH2/UBE2H (Accession#: P62256; Met1-Leu183) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title