missing translation for 'onlineSavingsMsg'
Learn More

UBE2J1 Antibody (OTI3D1), DyLight 350, Novus Biologicals™

Product Code. 18161657 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18161657 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18161657 Supplier Novus Biologicals Supplier No. NBP274747UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

UBE2J1 Monoclonal specifically detects UBE2J1 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBE2J1
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone OTI3D1
Conjugate DyLight 350
Dilution Western Blot, Flow Cytometry, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CGI-76, EC 6.3.2.19, HSPC153, HSPC205, HSU93243, HsUBC6e, MGC12555, NCUBE-1, NCUBE1HSUBC6e, Non-canonical ubiquitin-conjugating enzyme 1, non-canonical ubquitin conjugating enzyme 1, Ubc6p, ubiquitin-conjugating enzyme E2 J1, ubiquitin-conjugating enzyme E2, J1 (UBC6 homolog, yeast), Yeast ubiquitin-conjugating enzyme UBC6 homolog E
Gene Symbols UBE2J1
Host Species Mouse
Immunogen Full length recombinant protein of human UBE2J1 (NP_057105) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 51465.0
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.