missing translation for 'onlineSavingsMsg'
Learn More

UBE2R2 Antibody, Novus Biologicals™

Product Code. 18170072 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18170072 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18170072 Supplier Novus Biologicals Supplier No. NBP215154

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

UBE2R2 Polyclonal specifically detects UBE2R2 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBE2R2
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.69mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:10000, Immunocytochemistry/Immunofluorescence 1:100-1:1000
Formulation PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q712K3
Gene Alias CDC34BEC 6.3.2.19, E2-CDC34B, FLJ20419, MGC10481, UBC3BUbiquitin-protein ligase R2, Ubiquitin carrier protein R2, ubiquitin-conjugating enzyme E2 R2, Ubiquitin-conjugating enzyme E2-CDC34B, ubiquitin-conjugating enzyme E2R 2, ubiquitin-conjugating enzyme UBC3B
Gene Symbols UBE2R2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human UBC3B. The exact sequence is proprietary.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54926
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.