missing translation for 'onlineSavingsMsg'
Learn More

UBE2T Antibody [Janelia Fluor« 669], Novus Biologicals Biologicals™

Product Code. 30510268 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30510268 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30510268 Supplier Novus Biologicals Supplier No. NBP338343JF669

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

UBE2T Polyclonal antibody specifically detects UBE2T in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBE2T
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 669
Formulation 50mM Sodium Borate
Gene Alias Cell proliferation-inducing gene 50 protein, EC 6.3.2.19, HSPC150, HSPC150 protein similar to ubiquitin-conjugating enzyme, PIG50, Ubiquitin carrier protein T, ubiquitin conjugating enzyme, ubiquitin-conjugating enzyme E2 T, ubiquitin-conjugating enzyme E2T (putative), Ubiquitin-protein ligase T
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-197 of human UBE2T (NP_054895.1).,, Sequence:, MQRASRLKRELHMLATEPPPGITCWQDKDQMDDLRAQILGGANTPYEKGVFKLEVIIPERYPFEPPQIRFLTPIYHPNIDSAGRICLDVLKLPPKGAWRPSLNIATVLTSIQLLMSEPNPDDPLMADISSEFKYNKPAFLKNARQWTEKHARQKQKADEEEMLDNLPEAGDSRVHNSTQKRKASQLVGIEKKFHPDV
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29089
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.