missing translation for 'onlineSavingsMsg'
Learn More

UBE4B Antibody, Allophycocyanin, R&D Systems™

Product Code. 18601546 Shop All Bio Techne Products
Change view
Click to view available options
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
18601546 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18601546 Supplier Novus Biologicals Supplier No. NBP298954APC
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

UBE4B Polyclonal specifically detects UBE4B in Human samples. It is validated for Immunohistochemistry/Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBE4B
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate APC
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human UBE4B.
Purification Method Antigen and protein A Affinity-purified
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10277
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.