missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBXN2B Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
| Antigen | UBXN2B |
|---|---|
| Concentration | 1000 μg/mL |
| Applications | Western Blot, ELISA |
| Form | Liquid |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18059365
|
Proteintech
25141-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18037405
|
Proteintech
25141-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The UBXN2B antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human UBXN2B. This antibody recognizes human, mouse antigen. The UBXN2B antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| UBXN2B | |
| Western Blot, ELISA | |
| Rabbit | |
| 137886 | |
| IgG | |
| Store at -20°C. Stable for one year after shipment. |
| 1000 μg/mL | |
| Liquid | |
| Human, Mouse | |
| NSFL1 cofactor p37, p37, p97 cofactor p37, UBX domain protein 2B, UBXN2B | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts