missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Uroplakin III Antibody (SR2331), Novus Biologicals™

Product Code. 30041760 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30041760 100 μL 100µL
30041610 50 μL 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30041760 Supplier Novus Biologicals™ Supplier No. NBP327853100ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Uroplakin III Monoclonal antibody specifically detects Uroplakin III in Human samples. It is validated for Flow Cytometry,Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Uroplakin III
Applications Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone SR2331
Conjugate Unconjugated
Dilution Flow Cytometry 1:100, Immunohistochemistry 1:1000-1:500
Formulation PBS (pH 7.4), 150mM NaCl, 50% glycerol
Gene Alias MGC119178, UP3a, UPIII, UPIIIA, UPK3, UPK3A, uroplakin 3, uroplakin 3A, Uroplakin III, uroplakin3a, uroplakin-3a
Host Species Rabbit
Immunogen A synthesized peptide derived from human Uroplakin III (Uniprot #: O75631)
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 7380
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.