missing translation for 'onlineSavingsMsg'
Learn More

USP1 Antibody (8F7) [Alexa Fluor« 350], Novus Biologicals Biologicals™

Product Code. 30519697 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30519697 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30519697 Supplier Novus Biologicals Supplier No. NBP343060AF350

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

USP1 Monoclonal antibody specifically detects USP1 in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Especificaciones

Antigen USP1
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 8F7
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias Deubiquitinating enzyme 1, EC 3.1.2.15, EC 3.4.19.12, hUBP, ubiquitin carboxyl terminal hydrolase 1, ubiquitin carboxyl-terminal hydrolase 1, ubiquitin specific peptidase 1, ubiquitin specific processing protease 1, ubiquitin specific protease 1, ubiquitin thioesterase 1, Ubiquitin thiolesterase 1, Ubiquitin-specific-processing protease 1, UBP
Host Species Mouse
Immunogen ubiquitin specific peptidase 1 - (Uniprot# O94782)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7398
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.