missing translation for 'onlineSavingsMsg'
Learn More

USP22 Antibody - BSA Free, Novus Biologicals™

Product Code. 18707773 Shop All Bio Techne Products
Change view
Click to view available options
Unit Size:
0.1mL
25µL
Quantity:
0.025 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize Quantity
18707773 0.1mL -
18226454 25µL 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18707773 Supplier Novus Biologicals Supplier No. NBP149644

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 6 publications

USP22 Polyclonal specifically detects USP22 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.