missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
V2 Vasopressin R/AVPR2 Polyclonal antibody specifically detects V2 Vasopressin R/AVPR2 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | V2 Vasopressin R/AVPR2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:2000 |
| Formulation | PBS (pH 7.3), 50% glycerol |
| Gene Alias | ADHRMGC138386, Antidiuretic hormone receptor, arginine vasopressin receptor 2, AVPR V2, DI1, DIR3, DIRNDI, Renal-type arginine vasopressin receptor, V2RMGC126533, vasopressin V2 receptor |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 232-371 of human AVPR2 (NP_000045.1). IHASLVPGPSERPGGRRRGRRTGSPGEGAHVSAAVAKTVRMTLVIVVVYVLCWAPFFLVQLWAAWDPEAPLEGAPFVLLMLLASLNSCTNPWIYASFSSSVSSELRSLLCCARGRTPPSLGPQDESCTTASSSLAKDTSS |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?