missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VAChT/SLC18A3 Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB110-74764-0.025ML
This item is not returnable.
View return policy
Description
VAChT/SLC18A3 Polyclonal specifically detects VAChT/SLC18A3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| VAChT/SLC18A3 | |
| Polyclonal | |
| Unconjugated | |
| Whole antisera with PBS with No Preservative | |
| MGC12716, solute carrier family 18 (vesicular acetylcholine), member 3, Solute carrier family 18 member 3, solute carrier family 18, member 3, VAChT, VACHTvesicular acetylcholine transporter | |
| Goat | |
| Unpurified | |
| RUO | |
| 6572 | |
| Reconstitute with 0.1 ml sterilized water to desired concentration. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1:300-1:2000, Immunohistochemistry 1:300-1:2000, Immunohistochemistry-Paraffin 1:300-1:2000, Immunohistochemistry-Frozen 1:10-1:500 | |
| Q16572, O35304, Q62666 | |
| SLC18A3 | |
| A synthetic peptide as a part of human, mouse and rat VACHT conjugated to immunogenic carrier protein was used as the immunogen. Use in Immunohistochemistry-Frozen reported in scientific literature (PMID 24400086) | |
| 0.025 mL | |
| Primary | |
| VACHT. | |
| Mouse, Human, Rat | |
| Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction