missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VAChT/SLC18A3 Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 2 publications
228.00 € - 561.00 €
Specifications
| Antigen | VAChT/SLC18A3 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 1:300-1:2000, Immunohistochemistry 1:300-1:2000, Immunohistochemistry-Paraffin 1:300-1:2000, Immunohistochemistry-Frozen 1:10-1:500 |
| Applications | Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18010764
|
Novus Biologicals
NB110-74764-0.025ML |
0.025 mL |
228.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18711374
|
Novus Biologicals
NB110-74764 |
561.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
VAChT/SLC18A3 Polyclonal specifically detects VAChT/SLC18A3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| VAChT/SLC18A3 | |
| Western Blot 1:300-1:2000, Immunohistochemistry 1:300-1:2000, Immunohistochemistry-Paraffin 1:300-1:2000, Immunohistochemistry-Frozen 1:10-1:500 | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Q16572, O35304, Q62666 | |
| 6572 | |
| IgG | |
| Unpurified | |
| VACHT. |
| LYOPH | |
| Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Goat | |
| Mouse, Human, Rat | |
| MGC12716, solute carrier family 18 (vesicular acetylcholine), member 3, Solute carrier family 18 member 3, solute carrier family 18, member 3, VAChT, VACHTvesicular acetylcholine transporter | |
| SLC18A3 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title