missing translation for 'onlineSavingsMsg'
Learn More

Butyl 4-hydroxybenzoate-ring-13C6 solution, 10 μg/mL in acetone, Honeywell Fluka™

Product Code. 15910122 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15910122 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15910122 Supplier Honeywell Fluka Supplier No. R11105ML
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

10 μg/mL in acetone, analytical standard, reference material

Ensure you have the information you need to quickly start your analysis, and pass relevant audits:

  • High quality inorganic and organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
  • Free expert technical support
TRUSTED_SUSTAINABILITY

Specifications

Packaging Glass Bottle
CAS 1416711-53-9, 67-64-1
Density 0.79 g/cm3
Flash Point -17°C
Physical Form Liquid
Boiling Point 55°C to 57°C
Industry Type Parabenes
Quantity 5 mL
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.