missing translation for 'onlineSavingsMsg'
Learn More

Valine (L-Valine), LoGiCal

Product Code. 30460089
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30460089 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30460089 Supplier LoGiCal Supplier No. LGCFOR1455.00
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

Valine (L-Valine)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-Valine
CAS 72-18-4
Analyte or Component Names L-Valine
Shipping Condition Room Temperature
Molecular Formula C5 H11 N O2
Certifications/Compliance ISO 9001
Synonym Valine, (S)-2-amino-3-methylbutanoic acid, (+)-2-Amino-3-methylbutyric acid, (2 S)-2-Amino-3-methylbutanoic acid, (S)-2-Amino-3-methylbutanoic acid, (S)-2-Amino-3-methylbutyric acid, (S)-Valine, (S)-alpha-Amino-beta-methylbutyric acid, 2-Amino-3-methylbutanoic acid, 7: PN: US20090069547 PAGE: 10 claimed protein, 80: PN: EP2071334 SEQID: 89 claimed protein, 80: PN: WO2009077864 SEQID: 89 claimed protein, L-(+)-alpha-Aminoisovaleric acid, L-alpha-Amino-beta-methylbutyric acid, NSC 76038, Phenylalanine Impurity D
InChI Formula InChI=1 S/C5H11NO2/c1-3(2)4(6)5(7)8/h3-4 H,6H2,1-2H3,(H,7,8)/t4-/m0/s1
SMILES CC(C)[C@H](N)C(=O)O
IUPAC Name (2 S)-2-amino-3-methylbutanoic acid
Industry Type Forensics
Molecular Weight (g/mol) 117.15
Quantity 10 mg
Formula Weight 117.079 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.