missing translation for 'onlineSavingsMsg'
Learn More

Vardenafil Dihydrochloride 1.0 mg/ml in Methanol (as free base), LoGiCal

Product Code. 30459491
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30459491 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30459491 Supplier LoGiCal Supplier No. LGCAMP1619.0001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Vardenafil dihydrochloride
CAS 224789-15-5
Analyte or Component Names Vardenafil dihydrochloride
Shipping Condition Ice pack (-20°C)
Molecular Formula C23 H32 N6 O4 S . 2 Cl H
Certifications/Compliance ISO 17034
Synonym Imidazo[5,1-f][1,2,4]triazin-4(1H)-one, 2-[2-ethoxy-5-[(4-ethyl-1-piperazinyl)sulfonyl]phenyl]-5-methyl-7-propyl-, hydrochloride (1:2), Piperazine, 1-[[3-(1,4-dihydro-5-methyl-4-oxo-7-propylimidazo[5,1-f][1,2,4]triazin-2-yl)-4-ethoxyphenyl]sulfonyl]-4-ethyl-, dihydrochloride (9CI), Vardenafil dihydrochloride
InChI Formula InChI=1S/C23H32N6O4S.2ClH/c1-5-8-20-24-16(4)21-23(30)25-22(26-29(20)21)18-15-17(9-10-19(18)33-7-3)34(31,32)28-13-11-27(6-2)12-14-28;;/h9-10,15H,5-8,11-14H2,1-4H3,(H,25,26,30);2*1H
Solution Type Single Solution
SMILES Cl.Cl.CCCc1nc(C)c2C(=O)NC(=Nn12)c3cc(ccc3OCC)S(=O)(=O)N4CCN(CC)CC4
IUPAC Name 2-[2-ethoxy-5-(4-ethylpiperazin-1-yl)sulfonylphenyl]-5-methyl-7-propyl-3H-imidazo[5,1-f][1,2,4]triazin-4-one;dihydrochloride
Solvent or Matrix Methanol
Industry Type Food Testing
Molecular Weight (g/mol) 561.52
Quantity 1 mL
Formula Weight 560.1739 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.