Learn More
Invitrogen™ Varicella zoster Virus glycoprotein E Recombinant Rabbit Monoclonal Antibody (276)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA560398
 View more versions of this product
Product Code. 30535260
  • 169.00 € / 20µg
Description
Varicella zoster Virus glycoprotein E Recombinant Monoclonal Antibody for Western Blot, ELISA
Varicella Zoster Virus (VZV), a member of the human herpes virus family, causes two distinct clinical manifestations: childhood chickenpox(Varicella) and shingles (zoster). Varicella is the outcome of the primary infection with VZV, whereas, zoster is the result of VZV reactivation from latently infected sensory ganglia which occurs predominantly in aging and immunosuppressed individuals' virons are spherical and 150-200 nm in diameter. Its lipid envelope encloses the nucleocapsid of 162 capsomeres arranged in a hexagonal form. Its DNA is a single linear, double strand molecule, 125,000 nt long.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Varicella zoster Virus glycoprotein E
Recombinant Monoclonal
1 mg/mL
PBS with no preservative
Recombinant Varicella-zoster virus (VZV)/HHV-3 gE/glycoprotein E protein (Met1-Tyr538).
20 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
276
Unconjugated
Rabbit
Protein A
RUO
Virus