missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VAV2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-27078
This item is not returnable.
View return policy
Description
VAV2 Polyclonal specifically detects VAV2 in Human, Canine, Primate samples. It is validated for Western Blot.
Specifications
| VAV2 | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| guanine nucleotide exchange factor VAV2, oncogene VAV2, Protein vav-2, vav 2 guanine nucleotide exchange factor, vav 2 oncogene, VAV-2 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen for this VAV2 antibody is 100% homologous in human, dog, and chimpanzee 95% in mouse, Chinese hamster, mole rat, elephant, rhesus monkey, horse and guinea pig, 89% homolgous in panda, cow, zebrafish, opossum, platypus, cichlid fish, pig, and rat. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 0.5 mg/mL | |
| Western Blot 1-5ug/ml | |
| P52735 | |
| VAV2 | |
| A portion of amino acids 760-810 from human VAV2 was used as the immunogen for this antibody. | |
| 0.1 mg | |
| Signal Transduction | |
| 7410 | |
| Human, Canine, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction