missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VAV2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
213.00 € - 361.00 €
Specifications
| Antigen | VAV2 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 1-5ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18196354
|
Novus Biologicals
NBP2-27078SS |
0.025 mg |
213.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18106042
|
Novus Biologicals
NBP2-27078 |
0.1 mg |
361.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VAV2 Polyclonal specifically detects VAV2 in Human, Canine, Primate samples. It is validated for Western Blot.Specifications
| VAV2 | |
| Western Blot 1-5ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Canine, Primate | |
| P52735 | |
| 7410 | |
| A portion of amino acids 760-810 from human VAV2 was used as the immunogen for this antibody. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| guanine nucleotide exchange factor VAV2, oncogene VAV2, Protein vav-2, vav 2 guanine nucleotide exchange factor, vav 2 oncogene, VAV-2 | |
| VAV2 | |
| IgG | |
| Protein A purified | |
| The amino acid sequence used as immunogen for this VAV2 antibody is 100% homologous in human, dog, and chimpanzee 95% in mouse, Chinese hamster, mole rat, elephant, rhesus monkey, horse and guinea pig, 89% homolgous in panda, cow, zebrafish, opossum, platypus, cichlid fish, pig, and rat. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title