missing translation for 'onlineSavingsMsg'
Learn More

VEGF Antibody - BSA Free, Novus Biologicals™

Product Code. 18756683 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18756683 0.1 mL 0.1mL
18008643 0.025 mL 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18756683 Supplier Novus Biologicals Supplier No. NB1002381

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 5 publications

VEGF Polyclonal specifically detects VEGF in Human, Mouse, Rat, Bovine, Canine, Chicken, Equine, Guinea Pig samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VEGF
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 ug/ml, ELISA 1:500
Formulation PBS with 0.09% Sodium Azide
Gene Accession No. P15692
Gene Alias MVCD1, vascular endothelial growth factor A, Vascular permeability factor, VEGF-A, VEGFMGC70609, VPFvascular endothelial growth factor
Gene Symbols VEGFA
Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of the human VEGFA protein sequence (between residues 180-232). [Swiss-Prot: P15692]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cell Biology, HIF Target Genes, Hypoxia, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7422
Test Specificity Human, mouse, rat, horse, chicken, guinea pig, dog, and cow. Predicted to react with sheep and primate based on 100% sequence homology.
Target Species Human, Mouse, Rat, Bovine, Canine, Chicken, Equine, Guinea Pig
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.