missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEGF-B Antibody (SR1236), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22216-100ul
This item is not returnable.
View return policy
Description
VEGF-B Monoclonal antibody specifically detects VEGF-B in Human, Mouse samples. It is validated for Western Blot
Specifications
| VEGF-B | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| SR1236 | |
| Western Blot 1:500-1:2000 | |
| vascular endothelial growth factor B, VEGF-B, VEGF-related factor, VRFVEGFL | |
| A synthesized peptide derived from human VEGF-B (Uniprot #: P49765) | |
| 100 μg | |
| Angiogenesis, Cell Cycle and Replication | |
| 7423 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction