missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEGF-B Antibody (SR1236), Novus Biologicals™
Rabbit Monoclonal Antibody
338.00 € - 449.00 €
Specifications
| Antigen | VEGF-B |
|---|---|
| Clone | SR1236 |
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18621355
|
Novus Biologicals
NBP3-22216-100ul |
100 μg |
449.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18625174
|
Novus Biologicals
NBP3-22216-50ul |
50 μg |
338.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VEGF-B Monoclonal antibody specifically detects VEGF-B in Human, Mouse samples. It is validated for Western BlotSpecifications
| VEGF-B | |
| Western Blot 1:500-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| vascular endothelial growth factor B, VEGF-B, VEGF-related factor, VRFVEGFL | |
| A synthesized peptide derived from human VEGF-B (Uniprot #: P49765) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1236 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Cell Cycle and Replication | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 7423 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title