Learn More
VEGFR2/KDR/Flk-1 Antibody (89106R), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB3572RPEATT594
575.45 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
Product Code. 30036665
  • 719.00 € / 0.10mL
Description
VEGFR2/KDR/Flk-1 Monoclonal antibody specifically detects VEGFR2/KDR/Flk-1 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
VEGFR2/KDR/Flk-1
Monoclonal
PE/ATTO 594
PBS
Mouse
Protein A or G purified from cell culture supernatant
RUO
Primary
Human
Purified
Flow Cytometry
89106R
Flow Cytometry
CD309, CD309 antigen, EC 2.7.10, EC 2.7.10.1, Fetal liver kinase 1, fetal liver kinase-1, FLK-1, FLK1tyrosine kinase growth factor receptor, Kinase insert domain receptor, kinase insert domain receptor (a type III receptor tyrosine kinase), Protein-tyrosine kinase receptor flk-1, soluble VEGFR2, vascular endothelial growth factor receptor 2, VEGFR, VEGFR2, VEGFR-2
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human VEGFR2/KDR/Flk-1, Ala20-Glu764, Accession # P35968
0.1 mL
Angiogenesis, Apoptosis, Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, HIF Target Genes, Hypoxia, Phospho Specific, Signal Transduction, Stem Cell Markers, Tumor Suppressors, Tyrosine Kinases
3791
Store at 4C in the dark. Do not freeze.
IgG1