missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEPH1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
188.00 € - 470.00 €
Specifications
| Antigen | VEPH1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18618150
|
Novus Biologicals
NBP2-94600-0.02ml |
0.02 mL |
188.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18661691
|
Novus Biologicals
NBP2-94600-0.1ml |
0.1 mL |
470.00 €
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VEPH1 Polyclonal antibody specifically detects VEPH1 in Human, Mouse samples. It is validated for Western BlotSpecifications
| VEPH1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| FLJ12604, KIAA1692MGC111426, MELT, MGC126709, MGC142115, Protein melted, ventricular zone expressed PH domain homolog 1 (zebrafish), ventricular zone-expressed PH domain-containing protein homolog 1, VEPH | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 448-547 of human VEPH1 (NP_078897.2). KSLAFHTMLTKGVGSDDGEDENRGDIPASISLSEIDPLGQGNDKLPFKTDTERSQLGESSVSYPNIIHIDSENLSETVKENSQEETPETTASPIEYQDKL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 79674 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title