missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vesicular Stomatitis Indiana Virus Glycoprotein Antibody, FITC, R&D Systems™
Shop All Bio Techne ProductsDescription
Vesicular Stomatitis Indiana Virus Glycoprotein Polyclonal antibody specifically detects Vesicular Stomatitis Indiana Virus Glycoprotein in Virus samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Antigen | Vesicular Stomatitis Indiana Virus Glycoprotein |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | FITC |
| Dilution | Western Blot, ELISA |
| Formulation | PBS |
| Host Species | Rabbit |
| Immunogen | Rabbits immunized with purified, recombinant Vesicular Stomatitis Indiana Virus Glycoprotein (Accession#: NP_041715.1, Uniprot#: P03522, Met1-Ser467) |
| Purification Method | Antigen and protein A Affinity-purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?