missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vincamone, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
Vincamone
Specifications
Specifications
| Chemical Name or Material | Vincamone |
| CAS | 4880-88-0 |
| Analyte or Component Names | Vincamone |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H22 N2 O |
| Certifications/Compliance | ISO 17025 |
| Synonym | Desphosphoryltenofovir, (3alpha,16alpha)-Eburnamenin-14(15H)-one, Vincamone (7CI,8CI), 1H-Indolo[3,2,1-de]pyrido[3,2,1-ij][1,5]naphthyridine, eburnamenin-14(15H)-one deriv., (-)-Eburnamonine, (-)-Vincamone, (-)-cis-Eburnamonine, (3alpha,16alpha)-Eburnamonine, 16-Oxoeburnane, CH 846, Cervoxan, Eburnal Ritardo, Vinburnine, cis-Vincamone, l-Eburnamonine |
| SMILES | CC[C@@]12CCCN3CCc4c([C@H]13)n(C(=O)C2)c5ccccc45 |
| Molecular Weight (g/mol) | 294.38 |
| Quantity | 25 mg |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction