missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vinpocetine 1.0 mg/ml in Acetonitrile, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Vinpocetine |
| CAS | 42971-09-5 |
| Analyte or Component Names | Vinpocetine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C22 H26 N2 O2 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Vinpocetine, Ethyl (13aS,13bS)-13a-ethyl-2,3,5,6,13a,13b-hexahydro-1H-indolo[3,2,1-de]pyrido[3,2,1-ij][1,5]naphthyridine-12-carboxylate, 1H-Indolo[3,2,1-de]pyrido[3,2,1-ij][1,5]naphthyridine eburnamenine-14-carboxylic acid deriv., (+)-Apovincaminic acid ethyl ester, (+)-cis-Apovincaminic acid ethyl ester, AY 27255, Apovincaminic acid ethyl ester, Bravinton, Cavinton, Ceractin, Ethyl (+)-apovincaminate, Ethyl (+)-cis-apovincaminate, Ethyl apovincaminate, RGH 4405, TCV 3B, Ultra-Vinca, cis-Apovincaminic acid ethyl ester, (3alpha,16alpha)-Eburnamenine-14-carboxylic acid ethyl ester |
| InChI Formula | InChI=1S/C22H26N2O2/c1-3-22-11-7-12-23-13-10-16-15-8-5-6-9-17(15)24(19(16)20(22)23)18(14-22)21(25)26-4-2/h5-6,8-9,14,20H,3-4,7,10-13H2,1-2H3/t20-,22+/m1/s1 |
| Solution Type | Single Solution |
| SMILES | CCOC(=O)C1=C[C@]2(CC)CCCN3CCc4c([C@H]23)n1c5ccccc45 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?