missing translation for 'onlineSavingsMsg'
Learn More

Vinyl Acetate, Mikromol™

Product Code. 17972486
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17972486 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17972486 Supplier Mikromol Supplier No. MM0537.01
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Vinyl Acetate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Vinylacetate
CAS 108-05-4
Analyte or Component Names Vinylacetate
Shipping Condition Ice pack (-20°C)
Molecular Formula C4 H6 O2
Certifications/Compliance ISO 9001
Synonym Vinyl Acetate, Acetic acid vinyl ester (8CI), Ethenyl acetate, 1-Acetoxyethylene, Acetic acid, ethenyl ester, Acetoxyethene, Acetoxyethylene, NSC 8404, SN 12T, Vinyl A monomer, Vinyl acetate
InChI Formula InChI=1S/C4H6O2/c1-3-6-4(2)5/h3H,1H2,2H3
SMILES CC(=O)OC=C
IUPAC Name ethenyl acetate
Molecular Weight (g/mol) 86.09
Quantity 100 mg
Formula Weight 86.0368 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.