missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Viral MIP-III Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB698-SP
This item is not returnable.
View return policy
Description
MIP-III Monoclonal specifically detects MIP-III in Viral samples. It is validated for Western Blot.
Specifications
| MIP-III | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| Detects viral MIP-III in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant viral (rv) MIP-I, rvMIP-II, rvCMV UL146, or rvCMV UL147 is observed. | |
| Virus | |
| Purified |
| Western Blot | |
| 76621 | |
| Western Blot 1 ug/mL | |
| P88968 | |
| E. coli-derived recombinant human herpes virus-8 MIP-III Ser27-Pro114 Accession # P88968 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction