missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Viral MIP-III Antibody, R&D Systems™

Mouse Monoclonal Antibody
188.00 € - 496.00 €
Specifications
| Antigen | MIP-III |
|---|---|
| Clone | 76621 |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18753929
![]() |
R&D Systems
MAB698-SP |
25 μg |
188.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18149840
![]() |
R&D Systems
MAB698 |
500 μg |
496.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MIP-III Monoclonal specifically detects MIP-III in Viral samples. It is validated for Western Blot.Specifications
| MIP-III | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| E. coli-derived recombinant human herpes virus-8 MIP-III Ser27-Pro114 Accession # P88968 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects viral MIP-III in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant viral (rv) MIP-I, rvMIP-II, rvCMV UL146, or rvCMV UL147 is observed. |
| 76621 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Virus | |
| P88968 | |
| IgG1 | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title