missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Vitamin D BP Monoclonal antibody specifically detects Vitamin D BP in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein Array
Specifications
Specifications
| Antigen | Vitamin D BP |
| Applications | Immunohistochemistry (Paraffin), Protein Array |
| Classification | Monoclonal |
| Clone | VDBP/4481 |
| Conjugate | PerCP |
| Formulation | PBS |
| Gene Alias | DBP/GC, DBPVDBG, EC 6.3.1.5, Gc-globulin, GRD3, Group-specific component, group-specific component (vitamin D binding protein), hDBP, VDB, VDBP, vitamin D-binding alpha-globulin, vitamin D-binding protein |
| Host Species | Mouse |
| Immunogen | Recombinant fragment (around aa 35-175) of human Vitamin D BP protein (exact sequence is proprietary) (Uniprot: P02774) |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?